Learn More
Cayman Chemical Flavone, Purity: ≥98%, CAS: 525-82-6, 5 g
SDP
Catalog No. 502949918
Encompass_Preferred
A polyphenolic flavonoid; backbone scaffold of many pharmacologically active flavonoids; reduces proliferation of (EC50 = 54.8 µM), and induces apoptosis in, HT-29 colon cancer cells; enhances DENV-2 infectivity in Vero cells at 100 µg/ml
Catalog No. 50-294-9918 Supplier Cayman Chemical Supplier No. 340285 G
A polyphenolic flavonoid; backbone scaffold of many pharmacologically active flavonoids; reduces proliferation of (EC50 = 54.8 µM), and induces apoptosis in, HT-29 colon cancer cells; enhances DENV-2 infectivity in Vero cells at 100 µg/ml