Learn More
Cayman Chemical UnIdentIfIed PF1163A 2.5mg
SDP
Catalog No. 502987705
Encompass_Preferred
A depsipeptide antifungal; inhibits ERG synthesis (IC50 = 12 ng/ml); inhibits ERG25p, a C-4 methyl oxidase (MIC = 12.5 UG/ml); selectively inhibits growth of C. albicans (MIC = 8 UG/ml); acts synergistically with fluconazole to reduce growth of azole-resistant C. albicans (MIC = 0.0078 UG/ml); a more polar form of PF-1163B
Catalog No. 50-298-7705 Supplier Cayman Chemical Supplier No. 220652.5 MG
A depsipeptide antifungal; inhibits ERG synthesis (IC50 = 12 ng/ml); inhibits ERG25p, a C-4 methyl oxidase (MIC = 12.5 UG/ml); selectively inhibits growth of C. albicans (MIC = 8 UG/ml); acts synergistically with fluconazole to reduce growth of azole-resistant C. albicans (MIC = 0.0078 UG/ml); a more polar form of PF-1163B