Learn More
Cayman Chemical UnIdentIfIed PF1163B 2.5mg
SDP
Catalog No. 502987591
Encompass_Preferred
A depsipeptide antifungal; inhibits ERG synthesis (IC50 = 34 ng/ml); selectively inhibits growth of C. albicans (MIC = 32 UG/ml); acts synergistically with fluconazole to reduce growth of azole-resistant C. albicans (MIC = 0.0016 UG/ml); hydroxy form of PF-1163A
Catalog No. 50-298-7591 Supplier Cayman Chemical Supplier No. 220662.5 MG
Add to Cart
Add to Cart
A depsipeptide antifungal; inhibits ERG synthesis (IC50 = 34 ng/ml); selectively inhibits growth of C. albicans (MIC = 32 UG/ml); acts synergistically with fluconazole to reduce growth of azole-resistant C. albicans (MIC = 0.0016 UG/ml); hydroxy form of PF-1163A