Learn More
Cayman Chemical Santonin, Purity: ≥98%, CAS: 481-06-1, 5 g
SDP
Catalog No. 502941679
Encompass_Preferred
A sesquiterpenoid lactone; has been used as a building block in the synthesis of terpenoids; an anthelmintic that induces paralysis of A. lumbricoides
Catalog No. 50-294-1679 Supplier Cayman Chemical Supplier No. 274605 G
A sesquiterpenoid lactone; has been used as a building block in the synthesis of terpenoids; an anthelmintic that induces paralysis of A. lumbricoides