Description
- Intuitive design makes operation easy
- Visible needle utilizes the same technique as conventional pen needles
- Innovative push tab for one-handed activation of the safety mechanism
- Green cap for sterility and protection from the needle prior to injection
- Safety guard that covers the needle after injection to help prevent accidental needle sticks
- Dual safety indicator with visual and audible confirmation
- Both ends of the needle covered to protect both patient and healthcare professional
- Universal fit-compatible with most injection pens
Specifications
Specifications
| Needle Gauge | 30 G |
| For Use With (Application) | Drug Delivery Using Injection Pens |
| Length (Metric) | 5 mm |
| Product Type | Safety Pen Needle |
| Disposable | Single-use |
| Product Line | Unifine SafeControl |
| For Use With (Equipment) | Insulin Pens |
| Sterility | Sterile |