Learn More
Glass Expansion UniFit-08-4 Assembly (PKT 4)
SDP
Catalog No. 502702977
Encompass_Preferred
UniFit-08-4 Assembly (PKT 4). GazFit/UniFit drain fitting for 8mm OD HydraMist spray chamber drain and barbed end for soft walled tubing for peristaltic pump.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2977 Supplier Glass Expansion Supplier No. GAZ084
UniFit-08-4 Assembly (PKT 4). GazFit/UniFit drain fitting for 8mm OD HydraMist spray chamber drain and barbed end for soft walled tubing for peristaltic pump.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001