Learn More
Glass Expansion UniFit Connector with 1/16 OD X 0.50mm ID x 700mm long sample tube (PKT. 10)
SDP
Catalog No. 502702959
Encompass_Preferred
UniFit Connector with 1/16 OD X 0.50mm ID x 700mm long sample tube. Package of 10 replacement sample lines. Compatible with Glass Expansion U-Series and DC Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For UniFit Connector with 0.5mm ID, 1.3mm OD x 700mm long sample tube, NC1625341. For UniFit Connector with 0.18mm ID, 1.3mm OD x 700mm long sample tube, NC1973499.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2959 Supplier Glass Expansion Supplier No. NFT1650
UniFit Connector with 1/16 OD X 0.50mm ID x 700mm long sample tube. Package of 10 replacement sample lines. Compatible with Glass Expansion U-Series and DC Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For UniFit Connector with 0.5mm ID, 1.3mm OD x 700mm long sample tube, NC1625341. For UniFit Connector with 0.18mm ID, 1.3mm OD x 700mm long sample tube, NC1973499.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001