Learn More
Glass Expansion UniFit Cap (PKT 10)
SDP
Catalog No. 502703019
Encompass_Preferred
UniFit Cap (PKT 10). Replacement UniFit caps for HydraMist spray chamber reductant/sample port.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3019 Supplier Glass Expansion Supplier No. 708032078
UniFit Cap (PKT 10). Replacement UniFit caps for HydraMist spray chamber reductant/sample port.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001