Learn More
Perkin Elmer US LLC UniFit Connector for SeaSpray and MicroMist Nebulizers - 0.75 mm I.D. x 1.3 mm O.D. x 700 mm Long Sample Tube, Qty. 10
Supplier:  Perkin Elmer US LLC N0777512
Encompass_Preferred
This assembly includes 10 UniFit connectors. It is included in the SeaSpray Nebulizer, 2 mL/min (N0775345), Conikal Nebulizer, 2 mL/min (N0777487), and Slurry Nebulizer, 4 mL/min (N0777488) for Optima 4x00/5x00/7x00/8x00 and Avio 200/500.
Catalog No. 50-210-5261