Learn More
Perkin Elmer US LLC UniFit Connector for SeaSpray and MicroMist Nebulizers - 0.50 mm I.D. x 1.3 mm O.D. x 700 mm Long Sample Tube, Qty. 10
Catalog No. 502105286
Encompass_Preferred
This assembly includes 10 UniFit connectors. It is included in the MicroMist Nebulizer, 0.6 mL/min (N0775346) and the MicroMist Nebulizer, 0.2 mL/min (N0775347) for Optima 4x00/5x00/7x00/8x00 and Avio 200/500.
Catalog No. 50-210-5286 Supplier Perkin Elmer US LLC Supplier No. N0777513
This assembly includes 10 UniFit connectors. It is included in the MicroMist Nebulizer, 0.6 mL/min (N0775346) and the MicroMist Nebulizer, 0.2 mL/min (N0775347) for Optima 4x00/5x00/7x00/8x00 and Avio 200/500.