Learn More
Research Products International Corp UniFit Pipet Tips, Non-Sterile, Yellow, 1 - 200 ul, Bulk, 1000 per Case
SDP
Catalog No. 50212212
Encompass
Yellow Pipet Tips fit all popular brands of pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA, Volac, Octapette and Titertek. Yellow tips have a unique beveled end that minimizes fluid retention and improves accuracy.
Available in a fully enclosed bag that protects tips from airborne contamination.
Autoclavable
Polypropylene
RNase, DNase free
Catalog No. 50-212-212 Supplier Research Products International Corp Supplier No. 141152
Yellow Pipet Tips fit all popular brands of pipettors including Pipetman, Eppendorf, Finnpipette, Socorex, MLA, Volac, Octapette and Titertek. Yellow tips have a unique beveled end that minimizes fluid retention and improves accuracy.
Available in a fully enclosed bag that protects tips from airborne contamination.
Autoclavable
Polypropylene
RNase, DNase free