Learn More
Glass Expansion UniFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10)
SDP
Catalog No. 502702958
Encompass_Preferred
UniFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10). Pack of 10 replacement sample lines to connect Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179) to your Glass Expansion U-Series or Direct Connect “DC” Series nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2958 Supplier Glass Expansion Supplier No. NFT1650300
UniFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10). Pack of 10 replacement sample lines to connect Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179) to your Glass Expansion U-Series or Direct Connect “DC” Series nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001