Learn More
Glass Expansion UniFit with 1/16 OD X 0.75mm ID x 700mm long sample tube (PKT. 3).
SDP
Catalog No. NC1943061
Encompass_Preferred
UniFit Connector with 0.75mm ID x 1.6mm (1/16”) OD x 700mm long tubing sample tube (PKT 3). Package of 3 replacement reductant inlet port lines for Glass Expansion’s HydraMist spray chamber. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1943061 Supplier Glass Expansion Supplier No. UFT1675
UniFit Connector with 0.75mm ID x 1.6mm (1/16”) OD x 700mm long tubing sample tube (PKT 3). Package of 3 replacement reductant inlet port lines for Glass Expansion’s HydraMist spray chamber. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001