Learn More
Perkin Elmer US LLC UniFit-EzyFit Sample Tube - qty. 6
Catalog No. 502104956
Encompass_Preferred
This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).
Catalog No. 50-210-4956 Supplier Perkin Elmer US LLC Supplier No. N0777061
This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).