Learn More
World Precision Instrument Uninsulated Mini Banana Plug
SDP
Catalog No. 502521494
Encompass
Common connector for WPI amplifiers
Banana plugs are often used for making connections with WPI amplifiers. This is a mini, uninsulated, male banana plug connector.  The jack and plug are gold-plated to resist corrosion and provide optimal electrical connectivity.
Catalog No. 50-252-1494 Supplier World Precision Instrument Supplier No. 2035
Common connector for WPI amplifiers
Banana plugs are often used for making connections with WPI amplifiers. This is a mini, uninsulated, male banana plug connector.  The jack and plug are gold-plated to resist corrosion and provide optimal electrical connectivity.