Learn More
IDEX Health & Science Upchurch UNION 1/4-28IN
Catalog No. NC1919066
Encompass_Preferred
Idex ZDV Low-Pressure Delrin Acetal Unions, White, Temperature Max: 60 deg.C.
Catalog No. NC1919066 Supplier IDEX Health & Science Upchurch Supplier No. P603
Idex ZDV Low-Pressure Delrin Acetal Unions, White, Temperature Max: 60 deg.C.