Learn More
Waters Corp Waters Union, 0.040
Supplier: Waters Corp WAT089801
This union is a spare part designed to connect T107 or T107 HF tubing assemblies and prep flow cells. It is particularly useful for applications with flow rates above 100 mL/min when connecting specific tubing assemblies from a prep column outlet.
- Connects tubing assemblies and prep flow cells.
- Supports applications with flow rates exceeding 100 mL/min.
- Supplied as a single unit per package.
- Replacement is recommended as required.