Learn More
Trajan Scientific VICI Valco Union, Cheminert, 1/16" Tubing OD, 0.75mm Bore, PEEK, Includes Flangeless 1/4-28 Fittings For Tubing
Catalog No. 501090243
Encompass
VICI Valco Union, Cheminert, 1/16" Tubing OD, 0.75mm Bore, PEEK, Includes Flangeless 1/4-28 Fittings For Tubing. Made in the USA.
Catalog No. 50-109-0243 Supplier Trajan Scientific Supplier No. CUMPK
VICI Valco Union, Cheminert, 1/16" Tubing OD, 0.75mm Bore, PEEK, Includes Flangeless 1/4-28 Fittings For Tubing. Made in the USA.