Learn More
Microsolv Technology Corporation Union, Connector, Low Pressure, with 1/4-28 Screw Threads. Nylon. 1 EA.
Supplier:  Microsolv Technology Corporation 4530203
Encompass_Preferred
This "Union" is used to connect the same size low pressure tubing FEP ,PFA or PTFE on both ends, in the high pressures zones of HPLC systems. Due to its interior design, you can use this with 1/8" or 1/16th OD Tubing. The tubing ends will butt up against each other forming a zero dead volume connection. Nylon provides high strength and stiffness allowing for good connections when used with proper tubing connectors and all low pressure tubing grades.
Catalog No. 50-227-1435