Learn More
Microsolv Technology Corporation Union, Connector, Cross, High Pressure, 0.50mm Through Holes with 10-32 Screw Threads. Carbon PEEK. Use with 1/16" Tubing. 1 EA.
Catalog No. 502271453
Encompass_Preferred
This "Union" is used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
This item is not returnable. View return policy
Catalog No. 50-227-1453 Supplier Microsolv Technology Corporation Supplier No. 4730225
This "Union" is used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.