Learn More
Microsolv Technology Corporation Union, Connector, Cross, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Round with an Internal X. Use with 1/16" Tubing. 1 EA.
Catalog No. 502271529
Encompass_Preferred
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one.Use with Two Finger Tighten Connectors (nuts).
This item is not returnable. View return policy
Catalog No. 50-227-1529 Supplier Microsolv Technology Corporation Supplier No. 49302CX10
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one.Use with Two Finger Tighten Connectors (nuts).