Learn More
Microsolv Technology Corporation Union, Connector Kit, Cross, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Round with an Internal X. Use with 1/16" Tubing. 1 EA.
Catalog No. 502271530
Encompass_Preferred
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Supplied with Four Finger Tighten Connectors (nuts).
This item is not returnable. View return policy
Catalog No. 50-227-1530 Supplier Microsolv Technology Corporation Supplier No. 49302CX11
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Supplied with Four Finger Tighten Connectors (nuts).