Learn More
Microsolv Technology Corporation Union, Connector Kits, Cross, High Pressure, 0.50mm Through Holes with 10-32 Screw Threads. Carbon PEEK, with Connector Nuts. Use with 1/16" Tubing. 2/PK.
Catalog No. 502271454
Encompass_Preferred
These "Union" Kits are used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Supplied with 2 Cross Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
This item is not returnable. View return policy
Catalog No. 50-227-1454 Supplier Microsolv Technology Corporation Supplier No. 4730226
These "Union" Kits are used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Cross Connection" is often used to split flow into two or three directions or to combine two or three streams into one. Supplied with 2 Cross Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.