Learn More
Microsolv Technology Corporation Union, Connector Kits, Tee, High Pressure, 0.50mm Through Holes with 10-32 Screw Threads. Carbon PEEK, with Connector Nuts. Use with 1/16" Tubing. 2/PK.
Catalog No. 502271452
Encompass_Preferred
These "Union" Tee's are used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Tee Connection" is often used to split flow into two directions or to combine two streams into one. Supplied with 2 Tee Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
This item is not returnable. View return policy
Catalog No. 50-227-1452 Supplier Microsolv Technology Corporation Supplier No. 4730212
These "Union" Tee's are used to connect the same size PEEK or Stainless Steel tubing on all ends, in the high pressures zones of HPLC systems. A "Tee Connection" is often used to split flow into two directions or to combine two streams into one. Supplied with 2 Tee Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.