Learn More
Microsolv Technology Corporation Union, Connector Kit, High Pressure, 0.020" Through Hole with 10-32 Screw Threads. Stainless Steel, Straight Line. Use with 1/16" Tubing. 1 EA.
Catalog No. 502270857
Encompass_Preferred
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. Supplied with Two Stainless Steel Wrench Tighten Connectors and matching Ferrules.
This item is not returnable. View return policy
Catalog No. 50-227-0857 Supplier Microsolv Technology Corporation Supplier No. 4940202
This low dead volume, "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. Supplied with Two Stainless Steel Wrench Tighten Connectors and matching Ferrules.