Learn More
Microsolv Technology Corporation Union, Connector Kits, High Pressure, 0.50mm Through Hole with 10-32 Screw Threads. Carbon PEEK, Straight Line with Connector Nuts. Use with 1/16" Tubing. 2/PK.
Catalog No. 502271450
Encompass_Preferred
This "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. Supplied with 2 Straight Line Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.
This item is not returnable. View return policy
Catalog No. 50-227-1450 Supplier Microsolv Technology Corporation Supplier No. 4730202
This "Union" is used to connect the same size PEEK or Stainless Steel tubing on both ends, in the high pressures zones of HPLC systems. Supplied with 2 Straight Line Unions and Four Finger Tightened Connectors (nuts). Carbon PEEK has been reinforced with carbon fiber, giving it the highest strength and stiffness of all the PEEK grades and improving its load-carrying capacity.