Learn More
Grainger Union Elbow 1/2 in PK10
Catalog No. 502763774
Encompass_Preferred
The Parker 16W611 Union Elbow is a composite union elbow fitting for use in fluid power systems. It has a maximum pressure rating of 3000 psi and temperature range of -40°F to 250°F. The elbow has a 1/4" NPT female thread on both ends and is made of glass reinforced nylon composite material. This lightweight elbow provides corrosion resistance and has vibration damping properties. With its 360 degree orientation, it allows flexibility in installation. The elbow dimensions are 1-1/4" x 1-3/4" x 1-1/4". This composite elbow by Parker is designed for use in pneumatic applications.
Catalog No. 50-276-3774 Supplier Grainger Supplier No. 16W611
The Parker 16W611 Union Elbow is a composite union elbow fitting for use in fluid power systems. It has a maximum pressure rating of 3000 psi and temperature range of -40°F to 250°F. The elbow has a 1/4" NPT female thread on both ends and is made of glass reinforced nylon composite material. This lightweight elbow provides corrosion resistance and has vibration damping properties. With its 360 degree orientation, it allows flexibility in installation. The elbow dimensions are 1-1/4" x 1-3/4" x 1-1/4". This composite elbow by Parker is designed for use in pneumatic applications.