Learn More
Grainger Union Elbow 1/4 in PK10
Catalog No. 502764389
Encompass_Preferred
The Parker 16W609 Union Elbow is a composite union elbow fitting for use in fluid power systems. It has a maximum pressure rating of 5000 psi and temperature range of -65 to 250°F. The body material is glass reinforced nylon composite for corrosion resistance. It has a 1/4" NPT female thread on both ends. The elbow allows a 90 degree turn in tubing or hose routing. This composite elbow provides an economical, lightweight option for low pressure pneumatic applications. With proper installation and operating within pressure and temperature limits, it offers durable leak-free performance.
Catalog No. 50-276-4389 Supplier Grainger Supplier No. 16W609
The Parker 16W609 Union Elbow is a composite union elbow fitting for use in fluid power systems. It has a maximum pressure rating of 5000 psi and temperature range of -65 to 250°F. The body material is glass reinforced nylon composite for corrosion resistance. It has a 1/4" NPT female thread on both ends. The elbow allows a 90 degree turn in tubing or hose routing. This composite elbow provides an economical, lightweight option for low pressure pneumatic applications. With proper installation and operating within pressure and temperature limits, it offers durable leak-free performance.