Learn More
Trajan Scientific Union Medical Life Science Lab 1/EA
SDP
Catalog No. 5000494442
Encompass_Preferred
Glass lined stainless steel bulkhead union with 0 8 mm ID (no restriction)
Catalog No. 50-004-94442 Supplier Trajan Scientific Supplier No. 1236306
Glass lined stainless steel bulkhead union with 0 8 mm ID (no restriction)