Learn More
Hach Company Union Mt. Hardware SS for 3700
Catalog No. 5000207082 Shop All Hach Company Products
Encompass
Union Mounting Hardware for 3700 Series316 stainless steel union adapter and standard 2-inch pipe tee for use with 3700-series inductive/electrodeless conductivity sensors
Catalog No. 50-002-07082 Supplier Hach Company Supplier No. MH518N3NZ
Union Mounting Hardware for 3700 Series316 stainless steel union adapter and standard 2-inch pipe tee for use with 3700-series inductive/electrodeless conductivity sensors