Learn More
Hach Company Union Mt. Hardware SS for 3700
Catalog No. 5000207232 Shop All Hach Company Products
Encompass
Union Mounting Hardware for 3700-series SensorsCPVC union adapter and standard 2-inch pipe tee for use with the 3700-series inductive/electrodeless conductivity sensors
Catalog No. 50-002-07232 Supplier Hach Company Supplier No. MH538N3NZ
Union Mounting Hardware for 3700-series SensorsCPVC union adapter and standard 2-inch pipe tee for use with the 3700-series inductive/electrodeless conductivity sensors