Learn More
Hach Company Union Mt. Hardware SS w/o Tee
Catalog No. 5000207060 Shop All Hach Company Products
Encompass
Union Mounting Hardware-316 SS without Tee 316 stainless steel union adapter No Tee for use with 3700-series inductive/electrodeless conductivity sensors
Catalog No. 50-002-07060 Supplier Hach Company Supplier No. MH518M3NZ
Union Mounting Hardware-316 SS without Tee 316 stainless steel union adapter No Tee for use with 3700-series inductive/electrodeless conductivity sensors