Learn More
Microsolv Technology Corporation Union Peek .010 Body Only
Catalog No. 5001451629
Encompass_Preferred
Union, Connector, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Straight Line. Use with 1/16th Tubing. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51629 Supplier Microsolv Technology Corporation Supplier No. 4930203
Union, Connector, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Straight Line. Use with 1/16th Tubing. MicroSolv Brand. 1/EA.