Learn More
Microsolv Technology Corporation Union Peek .010 Hole Complt
Catalog No. 5001451627
Encompass_Preferred
Union, Connector Kit, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Straight Line. Use with 1/16th Tubing. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51627 Supplier Microsolv Technology Corporation Supplier No. 4930201
Union, Connector Kit, High Pressure, 0.30mm Through Hole with 10-32 Screw Threads. PEEK, Straight Line. Use with 1/16th Tubing. MicroSolv Brand. 1/EA.