Learn More
IDEX Health & Science Upchurch UNION PEEK .050 THRU HOLE
Catalog No. NC1919099
Encompass_Preferred
Idex Peek Low-Pressure Union Assembly, Natural, For Use With: With 2 P-300/P-335, 1/8 in., 1/4-28, 0.050 in. (1.25 mm) THRU, PEEK, Pressure Rating: 1000 psi, Temperature Max: 125 deg.C
Catalog No. NC1919099 Supplier IDEX Health & Science Upchurch Supplier No. P703
Idex Peek Low-Pressure Union Assembly, Natural, For Use With: With 2 P-300/P-335, 1/8 in., 1/4-28, 0.050 in. (1.25 mm) THRU, PEEK, Pressure Rating: 1000 psi, Temperature Max: 125 deg.C