Learn More
IDEX Health & Science Upchurch UNION POLY 1/4-28IN
Catalog No. NC1919075
Encompass_Preferred
Idex Low-Pressure Unions: Polypropylene, Natural, Temperature Max: 40 deg.C
Catalog No. NC1919075 Supplier IDEX Health & Science Upchurch Supplier No. P620
Idex Low-Pressure Unions: Polypropylene, Natural, Temperature Max: 40 deg.C