Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin Fitting, PFA, Straight, Compression Reducer, 1/2"" OD x 3/8"" OD
Catalog No. 50631991
Encompass
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Catalog No. 50-631-991 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3132004
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.