Learn More
Trajan Scientific Union Restriction Medical Lab 1/EA
SDP
Catalog No. 5000493975
Encompass_Preferred
Glass lined stainless steel bulkhead union with 0 4 mm ID and 0 25 mm restriction at one end
Catalog No. 50-004-93975 Supplier Trajan Scientific Supplier No. 1236302
Glass lined stainless steel bulkhead union with 0 4 mm ID and 0 25 mm restriction at one end