Learn More
Trajan Scientific Union Restriction Medical Lab 1/EA
SDP
Supplier:  Trajan Scientific 1236300
Encompass_Preferred
Glass lined stainless steel bulkhead union with 0 5 mm ID and 0 35 mm restriction at one end
Catalog No. 50-004-93664