Learn More
Microsolv Technology Corporation Union Ss Ldv 0.010 Holew/Nut
Catalog No. 5001452546
Encompass_Preferred
Union, Connector Kit, High Pressure, 0.010"" Through Hole with 10-32 Screw Threads. Stainless Steel, Straight Line. Use with 1/16th Tubing. BASIK Brand. 1/EA.
Catalog No. 50-014-52546 Supplier Microsolv Technology Corporation Supplier No. 4940201
Union, Connector Kit, High Pressure, 0.010"" Through Hole with 10-32 Screw Threads. Stainless Steel, Straight Line. Use with 1/16th Tubing. BASIK Brand. 1/EA.