Learn More
Grainger Union Tee 3/8 in PK10
Catalog No. 502763750
Encompass_Preferred
The Parker 16W613 is a composite union tee fitting made from engineered composite material. It has a maximum pressure rating of 300 PSI at 73°F. This union tee has female NPT thread connections and is designed for use with air, water, oil and inert gas systems. The operating temperature range is -40°F to 180°F. Key features include corrosion resistance, vibration dampening, and lightweight. This composite fitting provides a reliable connection method while being an alternative to metal fittings in corrosive environments. With its specifications and capabilities, this union tee can be utilized in a wide variety of industrial applications.
Catalog No. 50-276-3750 Supplier Grainger Supplier No. 16W613
The Parker 16W613 is a composite union tee fitting made from engineered composite material. It has a maximum pressure rating of 300 PSI at 73°F. This union tee has female NPT thread connections and is designed for use with air, water, oil and inert gas systems. The operating temperature range is -40°F to 180°F. Key features include corrosion resistance, vibration dampening, and lightweight. This composite fitting provides a reliable connection method while being an alternative to metal fittings in corrosive environments. With its specifications and capabilities, this union tee can be utilized in a wide variety of industrial applications.