Learn More
Grainger Union Tee 3/8 x 3/8 x 5/32 in PK10
Catalog No. 502764798
Encompass_Preferred
The Parker 16W615 Union Tee is a composite union tee fitting made from engineered composite thermoplastic. It has a maximum pressure rating of 300 PSI at 73°F. This union tee has female NPT thread connections and is designed for use with air, water, oil and inert gas in industrial applications. The operating temperature range is 32°F to 180°F. It is lightweight, corrosion resistant and provides an economical alternative to metal fittings. The union tee has a black body with white lettering for easy identification. It is manufactured in the USA and is RoHS compliant. The dimensions are 3-1/2" x 2-5/8" x 2-5/8".
Catalog No. 50-276-4798 Supplier Grainger Supplier No. 16W615
The Parker 16W615 Union Tee is a composite union tee fitting made from engineered composite thermoplastic. It has a maximum pressure rating of 300 PSI at 73°F. This union tee has female NPT thread connections and is designed for use with air, water, oil and inert gas in industrial applications. The operating temperature range is 32°F to 180°F. It is lightweight, corrosion resistant and provides an economical alternative to metal fittings. The union tee has a black body with white lettering for easy identification. It is manufactured in the USA and is RoHS compliant. The dimensions are 3-1/2" x 2-5/8" x 2-5/8".