Learn More
IDEX Health & Science Upchurch UNION TEFZEL 1/4-28
Catalog No. NC1919083
Encompass_Preferred
Idex ZDW Tefzel Low-Pressure Unions, For Use With: Not Applicable, Natural, ETFE, Temperature Max: 80 deg.C
Catalog No. NC1919083 Supplier IDEX Health & Science Upchurch Supplier No. P623
Idex ZDW Tefzel Low-Pressure Unions, For Use With: Not Applicable, Natural, ETFE, Temperature Max: 80 deg.C