Learn More
IDEX Health & Science Upchurch UNION UPTIGHT TRUE ZDV .062
Catalog No. NC1919403
Encompass_Preferred
Upchurch Scientific High-Pressure Unions, For Use With: 1/16 in. O.D. Tubing, Stainless-steel, Diameter Inner: 0.67 in., Swept Volume: Near 0 uL
Catalog No. NC1919403 Supplier IDEX Health & Science Upchurch Supplier No. U438
Upchurch Scientific High-Pressure Unions, For Use With: 1/16 in. O.D. Tubing, Stainless-steel, Diameter Inner: 0.67 in., Swept Volume: Near 0 uL