Learn More
IDEX Health & Science Upchurch UNION UPTIGHT ZDV .020
Catalog No. NC1919391
Encompass_Preferred
Idex VHP Union Assembly, Diameter Outer: 0.5 mm, Swept Volume: 0.134 uL, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, Stainless Steel, Pressure Rating: 10000 psi
Catalog No. NC1919391 Supplier IDEX Health & Science Upchurch Supplier No. U402
Idex VHP Union Assembly, Diameter Outer: 0.5 mm, Swept Volume: 0.134 uL, Diameter Inner: 0.02 in., For Use With: 1/16 in. O.D. Tubing, Stainless Steel, Pressure Rating: 10000 psi