Learn More
Phenomenex Inc Union, ZDV, S.S, 0.010in (0.25mm) Thru-Hole
Catalog No. 502412872 Shop All Phenomenex Products
Encompass_Preferred
Union, ZDV, S.S, 0.010in (0.25mm) Thru-Hole for 1/16in OD Tubing, with Fittings
Catalog No. 50-241-2872 Supplier Phenomenex Inc Supplier No. U435
Union, ZDV, S.S, 0.010in (0.25mm) Thru-Hole for 1/16in OD Tubing, with Fittings