Learn More
IDEX Health & Science Upchurch Union Body PEEK .020 thru hole, for 1/16" OD"
Catalog No. NC1763493
Encompass_Preferred
Low pressure PEEK Union Body efficiently connects tubing in low pressure regions of a system where a dependable connection is imperative. Made from resilient PEEK it delivers excellent solvent resistance. It is designed for 1/16"OD Tubing, has a 1/4-28 Flat Bottom thread port style and comes in natural color.
Catalog No. NC1763493 Supplier IDEX Health & Science Upchurch Supplier No. P70201
Low pressure PEEK Union Body efficiently connects tubing in low pressure regions of a system where a dependable connection is imperative. Made from resilient PEEK it delivers excellent solvent resistance. It is designed for 1/16"OD Tubing, has a 1/4-28 Flat Bottom thread port style and comes in natural color.