Learn More
Microsolv Technology Corporation Unions Endure Body Only Hplc
Catalog No. 5001451519
Encompass_Preferred
Union, Connector, High Pressure, 0.50mm Through Hole with 10-32 Screw Threads. Carbon PEEK, Straight Line. Use with 1/16th Tubing. Endure Brand. 1/EA.
Catalog No. 50-014-51519 Supplier Microsolv Technology Corporation Supplier No. 4730201
Union, Connector, High Pressure, 0.50mm Through Hole with 10-32 Screw Threads. Carbon PEEK, Straight Line. Use with 1/16th Tubing. Endure Brand. 1/EA.