Learn More
Cole-Parmer Analtech 21521 Uniplates, 10x20cm/UV/Scored/Hard/Silica GHL/250um Thick/15um Part 25/Pk
Catalog No. NC1535054 Shop All Cole-Parmer Products
Encompass_Preferred
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
Catalog No. NC1535054 Supplier Cole-Parmer Supplier No. 3410228
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.