Learn More
Mitegen Uni-puck Starter Kit 1, Includes 7 Pucks, Shipping Cane, Dewar, Tongs & Tools for Cryogenic Sample Management
SDP
Catalog No. 501187202
Encompass
The Uni-puck Starter Kit 1 provides a comprehensive solution for efficient and secure cryogenic sample management, ideal for new setups or expanding existing workflows. This kit includes 7 Universal V1-Pucks for organized sample storage and transport. It is further equipped with 1 Shelved Puck Shipping Cane and 1 Double Puck Loading Dewar with Lid, ensuring safe and streamlined handling of pucks during shipping and transfers. For precise manipulation, the kit contains 1 Puck Wand, 1 Cane Lifting J-hook, 1 Puck Separator Tools set (including a base removal platform and porcupine push tool), 1 Puck Dewar Loading Tool, and 1 Bent Cryo-Tong. This versatile collection of tools and containers enables researchers to safely organize, grasp, position, load, and separate Uni-Pucks, minimizing handling risks and enhancing efficiency throughout the cryogenic sample preparation and analysis pipeline.
Catalog No. 50-118-7202 Supplier Mitegen Supplier No. MCPUPSK001
The Uni-puck Starter Kit 1 provides a comprehensive solution for efficient and secure cryogenic sample management, ideal for new setups or expanding existing workflows. This kit includes 7 Universal V1-Pucks for organized sample storage and transport. It is further equipped with 1 Shelved Puck Shipping Cane and 1 Double Puck Loading Dewar with Lid, ensuring safe and streamlined handling of pucks during shipping and transfers. For precise manipulation, the kit contains 1 Puck Wand, 1 Cane Lifting J-hook, 1 Puck Separator Tools set (including a base removal platform and porcupine push tool), 1 Puck Dewar Loading Tool, and 1 Bent Cryo-Tong. This versatile collection of tools and containers enables researchers to safely organize, grasp, position, load, and separate Uni-Pucks, minimizing handling risks and enhancing efficiency throughout the cryogenic sample preparation and analysis pipeline.